Novus Biologicals
Manufacturer Code:NBP266914
Catalog # NBP266914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 43161 |
Immunogen | Recombinant protein with human Junctophilin-2 N terminal |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: JP-2; JP2 junctophilin 2 junctophilin type 2 junctophilin-2; JP2NT; Junctophilin type 2; Junctophilin-2; Junctophilin-2 N-terminal fragment
Gene Aliases: CMH17; JP-2; JP2; JPH2
UniProt ID: (Human) Q9BR39
Entrez Gene ID: (Human) 57158
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.