Invitrogen
Manufacturer Code:PA520641
Catalog # PIPA520641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide near the carboxy terminus of human JPH2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMH17 JP-2 JP2 junctophilin type 2 junctophilin-2; JP-2; JP2NT; jph2 {ECO:0000312|RGD:1305196}; Junctophilin type 2; Junctophilin-2; Junctophilin-2 N-terminal fragment
Gene Aliases: 1110002E14Rik; CMH17; JP-2; JP2; JPH2
UniProt ID: (Human) Q9BR39, (Mouse) Q9ET78, (Rat) Q2PS20
Entrez Gene ID: (Human) 57158, (Mouse) 59091, (Rat) 296345
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.