Invitrogen
Manufacturer Code:PA520640
Catalog # PIPA520640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide near the carboxy terminus of human JPH1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: JP-1; JP-1 JP1 junctophilin type 1 junctophilin type1 junctophilin-1 mitsugumin72; Junctophilin type 1; Junctophilin-1; mitsugumin72
Gene Aliases: CMT2K; ENSMUSG00000054314; JP-1; JP1; JPH1
UniProt ID: (Human) Q9HDC5, (Mouse) Q9ET80
Entrez Gene ID: (Human) 56704, (Mouse) 57339, (Rat) 297748
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.