Invitrogen
Manufacturer Code:PA571125
Catalog # PIPA571125
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human JOSD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: josephin domain-containing 1; Josephin domain-containing protein 1; Josephin-1
Gene Aliases: 1300006C06Rik; dJ508I15.2; JOSD1; JSPH1; KIAA0063; mKIAA0063; RGD1305388; zgc:153682
UniProt ID: (Bovine) Q5EAE5, (Human) Q15040, (Rat) Q5BJY4, (Mouse) Q9DBJ6
Entrez Gene ID: (Dog) 481251, (Rabbit) 100341161, (Bovine) 510781, (Human) 9929, (Horse) 100055255, (Rat) 315134, (Mouse) 74158, (Guinea pig) 100729641, (Zebrafish) 767655
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.