Novus Biologicals
Manufacturer Code:NBP151544
Catalog # NBP151544
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10E4A4 |
Immunogen | Purified recombinant fragment of human MAPK10 (aa28-233) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: c-Jun N-terminal kinase 3; c-Jun N-terminal kinase 3 EC 2.7.11 EC 2.7.11.24 JNK3 alpha protein kinase JNK3A JNK3FLJ12099 MAP kinase 10 MAP kinase p49 3F12 MAPK 10 MGC50974 mitogen-activated protein kinase 10 p493F12 p54bSAPK PRKM10FLJ33785 stress activated protein kinase beta Stress-activated protein kinase JNK3; JNK3 alpha protein kinase; MAP kinase 10; MAP kinase p49 3F12; MAPK 10; Mitogen-activated protein kinase 10; SAPK1b; stress activated protein kinase beta; Stress-activated protein kinase 1b; Stress-activated protein kinase JNK3
Gene Aliases: JNK3; JNK3A; MAPK10; p493F12; p54bSAPK; PRKM10; SAPK1B
UniProt ID: (Human) P53779
Entrez Gene ID: (Human) 5602
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.