R&D Systems
Manufacturer Code:MAB1846
Catalog # MAB1846
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 252320 |
Immunogen | <EM>E. coli</EM>-derived recombinant human JNK2 isoform 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: c-Jun kinase 2; c-Jun kinase 2 c-Jun N-terminal kinase 2 EC 2.7.11 EC 2.7.11.24 JNK2A JNK2BETA JNK2JNK2ALPHA JNK-55 Jun kinase MAP kinase 9 MAPK 9 MAPK9 mitogen-activated protein kinase 9 p54a p54aSAPK PRKM9JNK2B SAPK SAPK1 alpha Stress-activated protein kinase JNK2; c-Jun N-terminal kinase 2; JNK-55; Jun kinase; MAP kinase 9; MAPK 9; Mitogen-activated protein kinase 9; SAPK1a; Stress-activated protein kinase 1a; Stress-activated protein kinase JNK2
Gene Aliases: JNK-55; JNK2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; MAPK9; p54a; p54aSAPK; PRKM9; SAPK; SAPK1A
UniProt ID: (Human) P45984
Entrez Gene ID: (Human) 5601
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.