Novus Biologicals
Manufacturer Code:NBP289621100UL
Catalog # NB020725
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 11 |
Immunogen | Recombinant Human JNK2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: c-Jun kinase 2; c-Jun kinase 2 c-Jun N-terminal kinase 2 EC 2.7.11 EC 2.7.11.24 JNK2A JNK2BETA JNK2JNK2ALPHA JNK-55 Jun kinase MAP kinase 9 MAPK 9 mitogen-activated protein kinase 9 p54a p54aSAPK PRKM9JNK2B SAPK Stress-activated protein kinase JNK2; c-Jun N-terminal kinase 2; JNK-55; Jun kinase; MAP kinase 9; MAPK 9; Mitogen-activated protein kinase 9; SAPK1a; Stress-activated protein kinase 1a; Stress-activated protein kinase JNK2
Gene Aliases: JNK-55; JNK2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; MAPK9; p54a; p54aSAPK; PRKM9; SAPK; SAPK1A
UniProt ID: (Human) P45984
Entrez Gene ID: (Human) 5601
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.