Novus Biologicals
Manufacturer Code:NB6001297
Catalog # NB6001297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 373-389 (KDQPSDAAVSSNATPSQ) of human JNK2 was used as immunogen GenBank no. gi 21237736 ref NP_002743.3 . This amino sequence is 100% homologous in cow and dog and 94% in mouse and rat. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-Jun kinase 2; c-Jun kinase 2 c-Jun N-terminal kinase 2 EC 2.7.11 EC 2.7.11.24 JNK2A JNK2BETA JNK2JNK2ALPHA JNK-55 Jun kinase MAP kinase 9 MAPK 9 mitogen-activated protein kinase 9 p54a p54aSAPK PRKM9JNK2B SAPK Stress-activated protein kinase JNK2; c-Jun N-terminal kinase 2; JNK-55; Jun kinase; MAP kinase 9; MAPK 9; Mitogen-activated protein kinase 9; SAPK1a; Stress-activated protein kinase 1a; Stress-activated protein kinase JNK2
Gene Aliases: JNK-55; JNK2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; MAPK9; p54a; p54aSAPK; PRKM9; SAPK; SAPK1A
UniProt ID: (Human) P45984
Entrez Gene ID: (Human) 5601
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.