Proteintech
Manufacturer Code:511531AP150UL
Catalog # 50-173-5920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | JNK2 Fusion Protein Ag1442 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c Jun N terminal kinase 2 JNK 55 JNK2 JNK2A JNK2ALPHA JNK2B JNK2BETA MAP kinase 9 MAPK 9 MAPK9 p54a p54aSAPK PRKM9 SAPK; c-Jun kinase 2; c-Jun N-terminal kinase 2; JNK-55; JNK/SAPK alpha; Jun kinase; MAP kinase 9; MAPK 9; mitogen activated protein kinase 9; Mitogen-activated protein kinase 9; p54-alpha; protein kinase, mitogen-activated 9; SAPK-alpha; SAPK1a; stress activated protein kinase alpha II; Stress-activated protein kinase 1a; Stress-activated protein kinase JNK2
Gene Aliases: AI851083; JNK-55; JNK2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; MAPK9; p54a; p54aSAPK; PRKM9; SAPK; SAPK1A
UniProt ID: (Human) P45984, (Rat) P49186, (Mouse) Q9WTU6
Entrez Gene ID: (Human) 5601, (Rat) 50658, (Mouse) 26420
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.