Novus Biologicals
Manufacturer Code:NBP222209
Catalog # NBP222209
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 100000 |
Immunogen | Purified recombinant fragment of human JNK1 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: c-Jun N-terminal kinase 1; c-Jun N-terminal kinase 1 EC 2.7.11 EC 2.7.11.24 JNK1 alpha protein kinase JNK1 beta protein kinase JNK1JNK1A2 JNK-46 JUN N-terminal kinase MAP kinase 8 MAPK 8 mitogen-activated protein kinase 8 mitogen-activated protein kinase 8 isoform JNK1 alpha1 mitogen-activated protein kinase 8 isoform JNK1 beta2 PRKM8JNK protein kinase JNK1 SAPK1JNK21B1/2 Stress-activated protein kinase 1 Stress-activated protein kinase JNK1; JNK-46; JUN N-terminal kinase; MAP kinase 8; Mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; SAPK1c; stress-activated protein kinase 1; Stress-activated protein kinase 1c; Stress-activated protein kinase JNK1
Gene Aliases: JNK; JNK-46; JNK1; JNK1A2; JNK21B1/2; MAPK8; PRKM8; SAPK1; SAPK1C
UniProt ID: (Human) Q15713
Entrez Gene ID: (Human) 5599
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.