Invitrogen
Manufacturer Code:PA517889
Catalog # PIPA517889
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit human JNK2 fusion protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-Jun N-terminal kinase 1; JNK-46; JNK-46 JNK1 alpha protein kinase JNK1 beta protein kinase JUN N-terminal kinase MAP kinase 8 MAPK 8 OTTHUMP00000019552 c-Jun N-terminal kinase 1 mitogen-activated protein kinase 8 isoform JNK1 alpha1 mitogen-activated protein kinase 8 isoform JNK1 beta2 protein kinase JNK1 stress-activated protein kinase 1 stress-activated protein kinase JNK1; JNK1 beta1 protein kinase; JUN N-terminal kinase; MAP kinase 8; MAPK 8; mitogen activated protein kinase 8; Mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; p54 gamma; protein kinase mitogen-activated 8; SAPK gamma; SAPK1c; stress-activated protein kinase 1; Stress-activated protein kinase 1c; Stress-activated protein kinase JNK1
Gene Aliases: AI849689; JNK; JNK-46; JNK1; jnk1a-1; JNK1A2; JNK21B1/2; MAPK8; PRKM8; SAPK1; SAPK1C; zgc:112379
UniProt ID: (Human) Q15713, (Rat) P49185, (Mouse) Q91Y86, (Zebrafish) Q9DGD9
Entrez Gene ID: (Human) 5599, (Bovine) 539941, (Chinese hamster) 100771734, (Rat) 116554, (Mouse) 26419, (Zebrafish) 65236
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.