Novus Biologicals
Manufacturer Code:NBP250457C
Catalog # NBP250457C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HMY117 |
Immunogen | Synthetic peptide from the c terminus of the peptide sequence identical for both isoform 1 and 2 |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ37870 junction mediating and regulatory protein p53 cofactor junction-mediating and regulatory protein junction-mediating and -regulatory protein MGC163496 WAS protein homology region 2 domain containing 1-like 3 WHDC1L3; Junction-mediating and -regulatory protein; junction-mediating and regulatory protein; WAS protein homology region 2 domain containing 1-like 3
Gene Aliases: JMY; WHAMM2; WHDC1L3
UniProt ID: (Human) Q8N9B5
Entrez Gene ID: (Human) 133746
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.