Invitrogen
Manufacturer Code:PA520809
Catalog # PIPA520809
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide from near the carboxy terminus of human JMJD7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bifunctional peptidase and (3S)-lysyl hydroxylase JMJD7; hCG&lowbar37726 jmjC domain-containing protein 7 jumonji domain-containing protein 7; JmjC domain-containing protein 7; Jumonji domain-containing protein 7; L-lysine (3S)-hydroxylase JMJD7
Gene Aliases: JMJD7
UniProt ID: (Human) P0C870, (Mouse) P0C872
Entrez Gene ID: (Human) 100137047, (Rat) 100137086, (Mouse) 433466
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.