Novus Biologicals
Manufacturer Code:NBP298745UV
Catalog # NB105463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human JMJD6/PSR. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bifunctional arginine demethylase and lysyl-hydroxylase JMJD6; bifunctional arginine demethylase and lysyl-hydroxylase JMJD6 EC 1.14.11 EC 1.14.11.- Histone arginine demethylase JMJD6 JmjC domain-containing protein 6 jumonji domain containing 6 Jumonji domain-containing protein 6 KIAA0585PSR Lysyl-hydroxylase JMJD6 Peptide-lysine 5-dioxygenase JMJD6 Phosphatidylserine receptorPTDSR Protein PTDSR PTDSR1; Histone arginine demethylase JMJD6; JmjC domain-containing protein 6; jumonji domain containing 6; Jumonji domain-containing protein 6; Lysyl-hydroxylase JMJD6; Peptide-lysine 5-dioxygenase JMJD6; Phosphatidylserine receptor; Protein PTDSR
Gene Aliases: JMJD6; KIAA0585; PSR; PTDSR; PTDSR1
UniProt ID: (Human) Q6NYC1
Entrez Gene ID: (Human) 23210
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.