Invitrogen
Manufacturer Code:PA522132
Catalog # PIPA522132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contain a sequence corresponding to a region within amino acids 1 and 234 (Q6NYC1) of PSR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apoptotic cell clearance receptor PtdSerR; Bifunctional arginine demethylase and lysyl-hydroxylase JMJD6; Histone arginine demethylase JMJD6; JmjC domain-containing protein 6; jumonji domain containing 6; Jumonji domain-containing protein 6; Lysyl-hydroxylase JMJD6; Peptide-lysine 5-dioxygenase JMJD6; Phosphatidylserine receptor; Protein PTDSR; PSR PTDSR PTDSR1 bifunctional arginine demethylase and lysyl-hydroxylase JMJD6 histone arginine demethylase JMJD6 jmjC domain-containing protein 6 jumonji domain-containing protein 6 lysyl-hydroxylase JMJD6 peptide-lysine 5-dioxygenase JMJD6 phosphatidylserine receptor
Gene Aliases: 5730436I23Rik; D11Ertd195e; JMJD6; KIAA0585; mKIAA0585; PSR; PtdSerR; PTDSR; PTDSR1
UniProt ID: (Human) Q6NYC1, (Mouse) Q9ERI5
Entrez Gene ID: (Human) 23210, (Mouse) 107817
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.