OriGene
Manufacturer Code:TA810952
Catalog # 50-168-3727
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5E8 |
Immunogen | Full length human recombit protein of human JMJD5 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Bifunctional peptidase and arginyl-hydroxylase JMJD5; JmjC domain-containing protein 5; JMJD5; Jumonji C domain-containing protein 5; jumonji domain containing 5; jumonji domain-containing protein 5; L-arginine (3R)-hydroxylase KDM8; lysine (K)-specific demethylase 8
Gene Aliases: JMJD5; KDM8
UniProt ID: (Human) Q8N371
Entrez Gene ID: (Human) 79831
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.