Invitrogen
Manufacturer Code:PA544862
Catalog # PIPA544862
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human JMJD5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: arginyl C3-hydroxylase KDM8; Bifunctional peptidase and arginyl-hydroxylase JMJD5; JmjC domain-containing protein 5; Jumonji C domain-containing protein 5; jumonji domain containing 5; jumonji domain-containing protein 5; L-arginine (3R)-hydroxylase KDM8; lysine (K)-specific demethylase 8; Lysine-specific demethylase 8
Gene Aliases: 3110005O21Rik; JMJD5; KDM8; RGD1304823; zgc:173863; zgc:91962
UniProt ID: (Bovine) Q1JP61, (Human) Q8N371, (Mouse) Q9CXT6, (Rat) Q497B8
Entrez Gene ID: (Horse) 100064574, (Bovine) 507905, (Human) 79831, (Dog) 608058, (Rabbit) 100358867, (Mouse) 77035, (Guinea pig) 100723684, (Rat) 308976, (Zebrafish) 436936
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.