Novus Biologicals
Manufacturer Code:NBP177074
Catalog # NBP177074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human JMJD5. The immunogen is located within amino acids 350 - 400 of JMJD5. Amino Acid Squence: TSQVDVENPDLEKFPKFA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bifunctional peptidase and arginyl-hydroxylase JMJD5; EC 1.14.11.27 FLJ13798 JmjC domain-containing protein 5 jumonji domain containing 5 Jumonji domain-containing protein 5 KDM8 lysine-specific demethylase 8; JmjC domain-containing protein 5; Jumonji C domain-containing protein 5; jumonji domain containing 5; jumonji domain-containing protein 5; L-arginine (3R)-hydroxylase KDM8; lysine (K)-specific demethylase 8
Gene Aliases: JMJD5; KDM8
UniProt ID: (Human) Q8N371
Entrez Gene ID: (Human) 79831
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.