Novus Biologicals
Manufacturer Code:NBP103357AF594
Catalog # NP10337A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to a C-terminal portion of the human JMJD2D protein (within residues 460-520). [Swiss-ProtNo. Q6B0I6] |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: [histone H3]-trimethyl-L-lysine(9) demethylase 4D; EC 1.14.11 EC 1.14.11.- FLJ10251 JHDM3D JmjC domain-containing histone demethylation protein 3D JMJD2Dlysine-specific demethylase 4D jumonji domain containing 2D Jumonji domain-containing protein 2D lysine (K)-specific demethylase 4D MGC141909; JmjC domain-containing histone demethylation protein 3D; jumonji domain containing 2D; Jumonji domain-containing protein 2D; lysine (K)-specific demethylase 4D; Lysine-specific demethylase 4D
Gene Aliases: JHDM3D; JMJD2D; KDM4D
UniProt ID: (Human) Q6B0I6
Entrez Gene ID: (Human) 55693
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.