Abnova
Manufacturer Code:H00055693K
Catalog # 89-207-525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human JMJD2D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human JMJD2D peptide using ARM Technology.
For Research Use Only
Protein Aliases: [histone H3]-trimethyl-L-lysine(9) demethylase 4D; FLJ10251 KDM4D MGC141909; JmjC domain-containing histone demethylation protein 3D; jumonji domain containing 2D; Jumonji domain-containing protein 2D; lysine (K)-specific demethylase 4D; Lysine-specific demethylase 4D
Gene Aliases: JHDM3D; JMJD2D; KDM4D
UniProt ID: (Human) Q6B0I6
Entrez Gene ID: (Human) 55693
Molecular Function: transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.