Invitrogen
Manufacturer Code:PA578433
Catalog # PIPA578433
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human KDM4C. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H3]-trimethyl-L-lysine(9) demethylase 4C; GASC-1 protein; GASC1 JHDM3C JMJD2C KDM4C KDM4C TDRD14C bA146B14.1 lysine demethylase 4C; Gene amplified in squamous cell carcinoma 1 protein; JmjC domain-containing histone demethylation protein 3C; Jumonji domain-containing protein 2C; lysine (K)-specific demethylase 4C; Lysine-specific demethylase 4C; tudor domain containing 14C
Gene Aliases: GASC1; JHDM3C; JMJD2C; KDM4C; KIAA0780; TDRD14C
UniProt ID: (Human) Q9H3R0
Entrez Gene ID: (Human) 23081
Molecular Function: transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.