Invitrogen
Manufacturer Code:PA5115459
Catalog # PIPA5115459
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KDM4A corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H3]-trimethyl-L-lysine(36) demethylase 4A; [histone H3]-trimethyl-L-lysine(9) demethylase 4A; D4Ertd222e JHDM3A JmjC domain-containing histone demethylation protein 3A Jmjd2 Jmjd2a jumonji C domain-containing histone demethylase 3A jumonji domain containing 2 jumonji domain containing 2A jumonji domain-containing protein 2A KDM4A Kiaa0677 lysine (K)-specific demethylase 4A lysine demethylase 4A lysine-specific demethylase 4A mKIAA0677 TDRD14A tudor domain containing 14A; JmjC domain-containing histone demethylation protein 3A; jumonji C domain-containing histone demethylase 3A; jumonji domain containing 2; jumonji domain containing 2A; Jumonji domain-containing protein 2A; lysine (K)-specific demethylase 4A; Lysine-specific demethylase 4A; tudor domain containing 14A
Gene Aliases: D4Ertd222e; JHDM3A; JMJD2; JMJD2A; KDM4A; KIAA0677; mKIAA0677; TDRD14A
UniProt ID: (Human) O75164, (Mouse) Q8BW72
Entrez Gene ID: (Human) 9682, (Rat) 313539, (Mouse) 230674
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.