Invitrogen
Manufacturer Code:PA520804
Catalog # PIPA520804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 20 amino acid peptide from near the amino terminus of human JMJD1C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Jumonji domain-containing protein 1C; Probable JmjC domain-containing histone demethylation protein 2C; RP11-10C13.2 TRIP8 TR-interacting protein 8 TRIP-8 jumonji domain-containing protein 1C probable JmjC domain-containing histone demethylation protein 2C thyroid hormone receptor interactor 8 thyroid receptor interacting protein 8 thyroid receptor-interacting protein 8; thyroid hormone receptor interactor 8; Thyroid receptor-interacting protein 8; TR-interacting protein 8
Gene Aliases: 5430433L24Rik; D630035I23Rik; JHDM2C; JMJD1C; Jmjdic; KIAA1380; Pr2; TRIP-8; TRIP8
UniProt ID: (Human) Q15652, (Mouse) Q69ZK6
Entrez Gene ID: (Human) 221037, (Mouse) 108829, (Rat) 171120
Molecular Function: transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.