Abnova
Manufacturer Code:H00221037A01
Catalog # 89-107-707
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 2 a.a. &bsim 99 a.a) partial recombinant protein with GST tag.,JMJD1C (NP_004232 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant JMJD1C.
For Research Use Only
Protein Aliases: DKFZp761F0118; FLJ14374; Jumonji domain-containing protein 1C; KIAA1380; Probable JmjC domain-containing histone demethylation protein 2C; RP11-10C13.2; thyroid hormone receptor interactor 8; Thyroid receptor-interacting protein 8; TR-interacting protein 8; TRIP8
Gene Aliases: JHDM2C; JMJD1C; KIAA1380; TRIP-8; TRIP8
UniProt ID: (Human) Q15652
Entrez Gene ID: (Human) 221037
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.