Invitrogen
Manufacturer Code:PA520803
Catalog # PIPA520803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 20 amino acid peptide from near the center of human JMJD1B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5qNCA C5orf7 JMJD1B NET22 jmjC domain-containing histone demethylation protein 2B jumonji domain containing 1B jumonji domain-containing protein 1B lysine-specific demethylase 3B nuclear protein 5qNCA; [histone H3]-dimethyl-L-lysine(9) demethylase 3B; JmjC domain-containing histone demethylation protein 2B; jumonji domain containing 1B; Jumonji domain-containing protein 1B; lysine (K)-specific demethylase 3B; Lysine-specific demethylase 3B; Nuclear protein 5qNCA
Gene Aliases: 5830462I21Rik; 5qNCA; C5orf7; JHDM2B; JMJD1B; KDM3B; KIAA1082; mKIAA1082; NET22
UniProt ID: (Human) Q7LBC6
Entrez Gene ID: (Human) 51780, (Mouse) 277250, (Rat) 682469
Molecular Function: transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.