Invitrogen
Manufacturer Code:PA551755
Catalog # PIPA551755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human JM4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Jena-Muenchen 4; JM4 Jena-Muenchen 4 PRA1 domain family 2 PRA1 family protein 2; PRA1 domain family 2; PRA1 domain family, member 2; PRA1 family protein 2
Gene Aliases: JM4; PRAF2; RGD1566287; Yip6a
UniProt ID: (Human) O60831
Entrez Gene ID: (Human) 11230, (Rat) 367743
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.