Invitrogen
Manufacturer Code:PA559182
Catalog # PIPA559182
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human JLP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-Jun NH2-terminal kinase-associated leucine zipper protein; C-Jun-amino-terminal kinase-interacting protein 4; C-jun-amino-terminal kinase-interacting protein 4 JNK-interacting protein 4 JIP-4 JNK-associated leucine-zipper protein JLP Sperm-associated antigen 9 Mitogen-activated protein kinase 8-interacting protein 4 Human lung cancer protein 6 HLC-6 Proliferation-inducing protein 6 Sperm-specific protein Sperm surface protein Protein highly expressed in testis PHET Sunday driver 1 Cancer/testis antigen 89 CT89 SPAG9 HSS KIAA0516 MAPK8IP4 SYD1; Cancer/testis antigen 89; CT89; HLC-6; Human lung cancer oncogene 6 protein; JIP-4; JLP; JNK interacting protein; JNK-associated leucine-zipper protein; JNK/SAPK-associated protein; lung cancer oncogene 4; Max-binding protein; Mitogen-activated protein kinase 8-interacting protein 4; PHET; proliferation-inducing gene 6; Proliferation-inducing protein 6; Protein highly expressed in testis; sperm associated antigen 9 variant 5; Sperm surface protein; Sperm-associated antigen 9; Sperm-specific protein; Sunday driver 1
Gene Aliases: CT89; HLC-6; HLC4; HLC6; HSS; JIP-4; JIP4; JLP; KIAA0516; MAPK8IP4; PHET; PIG6; SPAG9; SYD1
UniProt ID: (Human) O60271
Entrez Gene ID: (Human) 9043
Molecular Function: enzyme modulator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.