Invitrogen
Manufacturer Code:PA5100730
Catalog # PIPA5100730
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human JIP4 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3110018C07Rik 4733401I23Rik 4831406C20Rik AW552012 Cancer/testis antigen 89 c-Jun NH2-terminal kinase-associated leucine zipper protein C-Jun-amino-terminal kinase-interacting protein 4 CT89 HLC4 HLC6 HLC-6 HSS Human lung cancer oncogene 6 protein Jip4 JIP-4 JLP JNK interacting protein JNK/SAPK-associated protein JNK/SAPK-associated protein 2 JNK-associated leucine-zipper protein JNK-interacting leucine zipper JNK-interacting protein 4 JSAP2 JSAP2a Kiaa0516 lung cancer oncogene 4 Mapk8ip4 Max-binding protein mitogen-activated protein kinase 8-interacting protein 4 PHET PIG6 proliferation-inducing gene 6 Proliferation-inducing protein 6 Protein highly expressed in testis Spag9 sperm associated antigen 9 sperm associated antigen 9 variant 5 sperm surface protein Sperm-associated antigen 9 Sperm-specific protein Sunday driver 1 SYD1; c-Jun NH2-terminal kinase-associated leucine zipper protein; C-Jun-amino-terminal kinase-interacting protein 4; Cancer/testis antigen 89; CT89; HLC-6; Human lung cancer oncogene 6 protein; JIP-4; JLP; JNK interacting protein; JNK-associated leucine-zipper protein; JNK-interacting leucine zipper; JNK-interacting protein 4; JNK/SAPK-associated protein; JNK/SAPK-associated protein 2; JSAP2; lung cancer oncogene 4; Max-binding protein; Mitogen-activated protein kinase 8-interacting protein 4; PHET; proliferation-inducing gene 6; Proliferation-inducing protein 6; Protein highly expressed in testis; sperm associated antigen 9 variant 5; Sperm surface protein; Sperm-associated antigen 9; Sperm-specific protein; Sunday driver 1
Gene Aliases: 3110018C07Rik; 4733401I23Rik; 4831406C20Rik; AW552012; CT89; HLC-6; HLC4; HLC6; HSS; JIP-4; JIP4; JLP; JSAP2; JSAP2a; KIAA0516; MAPK8IP4; PHET; PIG6; SPAG9; SYD1
UniProt ID: (Human) O60271, (Mouse) Q58A65
Entrez Gene ID: (Human) 9043, (Mouse) 70834
Molecular Function:
enzyme modulator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.