Invitrogen
Manufacturer Code:PA536369
Catalog # PIPA536369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 615-664 of human JIP-3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-jun-amino-terminal kinase interacting protein 3; C-Jun-amino-terminal kinase-interacting protein 3; C-jun-amino-terminal kinase-interacting protein 3 JNK-interacting protein 3 JIP-3 JNK MAP kinase scaffold protein 3 Mitogen-activated protein kinase 8-interacting protein 3 JIP3 KIAA1066; homolog of Drosophila Sunday driver 2; JIP-3; JNK MAP kinase scaffold protein 3; JNK-interacting protein 3; JNK/SAPK-associated protein-1; JNK/stress-activated protein kinase-associated protein 1; Mitogen-activated protein kinase 8-interacting protein 3
Gene Aliases: JIP-3; JIP3; JSAP1; KIAA1066; MAPK8IP3; syd; SYD2
UniProt ID: (Human) Q9UPT6
Entrez Gene ID: (Human) 23162
Molecular Function: enzyme modulator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.