Invitrogen
Manufacturer Code:PA529245
Catalog # PIPA529245
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 529 and 804 of Human JIP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-Jun-amino-terminal kinase-interacting protein 2; homologous to mouse JIP-1; IB-2; IB2 JIP2 PRKM8IPL C-Jun-amino-terminal kinase-interacting protein 2 IB-2 JIP-2 JNK MAP kinase scaffold protein 2 JNK MAP kinase scaffold protein JIP2 JNK-interacting protein 2 homologous to mouse JIP-1 islet-brain 2 islet-brain-2 mitogen-activated protein kinase 8-interact; islet-brain 2; Islet-brain-2; JIP-2; JNK MAP kinase scaffold protein 2; JNK MAP kinase scaffold protein JIP2; JNK-interacting protein 2; mitogen activated protein kinase 8 interacting protein 2; Mitogen-activated protein kinase 8-interacting protein 2
Gene Aliases: 3230402N03Rik; AI847694; AW049958; IB-2; IB2; JIP2; MAPK8IP2; PRKM8IPL; R75148
UniProt ID: (Human) Q13387, (Mouse) Q9ERE9
Entrez Gene ID: (Human) 23542, (Mouse) 60597
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.