Novus Biologicals
Manufacturer Code:NBP131411
Catalog # NBP131411
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human JIP1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-Jun-amino-terminal kinase-interacting protein 2; C-Jun-amino-terminal kinase-interacting protein 2 homologous to mouse JIP-1 IB-2 IB2JIP2Mitogen-activated protein kinase 8-interacting protein 2 islet-brain 2 Islet-brain-2 JIP-2 JNK MAP kinase scaffold protein 2 JNK MAP kinase scaffold protein JIP2 JNK-interacting protein 2 mitogen-activated protein kinase 8 interacting protein 2 PRKM8 interacting protein-like PRKM8IPL; homologous to mouse JIP-1; IB-2; islet-brain 2; Islet-brain-2; JIP-2; JNK MAP kinase scaffold protein 2; JNK MAP kinase scaffold protein JIP2; JNK-interacting protein 2; Mitogen-activated protein kinase 8-interacting protein 2
Gene Aliases: IB-2; IB2; JIP2; MAPK8IP2; PRKM8IPL
UniProt ID: (Human) Q13387
Entrez Gene ID: (Human) 23542
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.