R&D Systems
Manufacturer Code:AF4366SP
Catalog # AF4366SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human JIP1 Met531-Asp666 Accession # Q6NUQ9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: C-Jun-amino-terminal kinase-interacting protein 1; IB-1; Islet-brain 1; JIP-1; JNK MAP kinase scaffold protein 1; JNK-interacting protein 1; Mitogen-activated protein kinase 8-interacting protein 1; PRKM8 interacting protein
Gene Aliases: IB1; JIP-1; JIP1; MAPK8IP1; PRKM8IP
UniProt ID: (Human) Q9UQF2
Entrez Gene ID: (Human) 9479
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.