Invitrogen
Manufacturer Code:PA578032
Catalog # PIPA578032
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human JARID2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: jumonji AT rich interactive domain 2 JMJ; jumonji homolog; jumonji, AT rich interactive domain 2; jumonji-like protein; Jumonji/ARID domain-containing protein 2; Protein Jumonji
Gene Aliases: JARID2; JMJ; jumonji
UniProt ID: (Human) Q92833, (Mouse) Q62315
Entrez Gene ID: (Human) 3720, (Mouse) 16468
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.