Abnova
Manufacturer Code:H00008284M03
Catalog # 89-105-826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4C6 |
Immunogen | JARID1D (NP_004644.2127 a.a. &bsim 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant JARID1D.
For Research Use Only
Protein Aliases: [histone H3]-trimethyl-L-lysine(4) demethylase 5D; H-Y; Histocompatibility Y antigen; Histone demethylase JARID1D; HY; HYA; Jumonji, AT rich interactive domain 1D (RBP2-like); Jumonji/ARID domain-containing protein 1D; KDM5D; KIAA0234; lysine (K)-specific demethylase 5D; Lysine-specific demethylase 5D; Protein SmcY; selected mouse cDNA on Y, human homolog of; SMC homolog, Y chromosome; SMCY; Smcy homolog, Y-linked
Gene Aliases: HY; HYA; JARID1D; KDM5D; KIAA0234; SMCY
UniProt ID: (Human) Q9BY66
Entrez Gene ID: (Human) 8284
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.