Abnova
Manufacturer Code:MAB11151
Catalog # 89-109-333
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to C-terminus of human JARID1C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant protein of JARID1C.
For Research Use Only
Protein Aliases: [histone H3]-trimethyl-L-lysine(4) demethylase 5C; DXS1272E; Histone demethylase JARID1C; JmjC domain-containing protein SMCX; Jumonji, AT rich interactive domain 1C (RBP2-like); Jumonji/ARID domain-containing protein 1C; KDM5C; lysine (K)-specific demethylase 5C; Lysine-specific demethylase 5C; MRXJ; MRXSJ; Protein SmcX; Protein Xe169; SMCX; Smcx homolog, X chromosome; Smcy homolog, X-linked; XE169
Gene Aliases: DXS1272E; JARID1C; KDM5C; MRX13; MRXJ; MRXSCJ; MRXSJ; SMCX; XE169
UniProt ID: (Human) P41229
Entrez Gene ID: (Human) 8242
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.