R&D Systems
Manufacturer Code:FAB7050A
Catalog # FAB7050A
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 209628 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse JAM-C (CD323) Val32-Asn241 Accession # Q9D8B7 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD323 JAM-2 JAM3 JAMC JAM-CFLJ14529 junctional adhesion molecule 3JAM-3 junctional adhesion molecule C; JAM-2; JAM-3; JAM-C; Junctional adhesion molecule 3; Junctional adhesion molecule C; sJAM-C; Soluble form of JAM-C
Gene Aliases: JAM-2; JAM-3; JAM-C; JAM3; JAMC; UNQ859/PRO1868
UniProt ID: (Human) Q9BX67
Entrez Gene ID: (Human) 83700
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.