Novus Biologicals
Manufacturer Code:NBP232343
Catalog # NBP232343
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This JAM3 antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide between 261-295 amino acids from the C-terminal region of human JAM3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: JAM-2; JAM-2 JAMC JAM-CFLJ14529 junctional adhesion molecule 3JAM-3 junctional adhesion molecule C; JAM-3; JAM-C; Junctional adhesion molecule 3; Junctional adhesion molecule C; sJAM-C; Soluble form of JAM-C
Gene Aliases: JAM-2; JAM-3; JAM-C; JAM3; JAMC; UNQ859/PRO1868
UniProt ID: (Human) Q9BX67
Entrez Gene ID: (Human) 83700
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.