R&D Systems
Manufacturer Code:AF1213
Catalog # AF1213
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse JAM-C Val32-Asn241 Accession # Q9D8B7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD323 JAM-2 JAM3 JAMC JAM-CFLJ14529 junctional adhesion molecule 3JAM-3 junctional adhesion molecule C; JAM-2; JAM-3; JAM-C; Junctional adhesion molecule 3; Junctional adhesion molecule C; sJAM-C; Soluble form of JAM-C
Gene Aliases: JAM-2; JAM-3; JAM-C; JAM3; JAMC; UNQ859/PRO1868
UniProt ID: (Human) Q9BX67
Entrez Gene ID: (Human) 83700
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.