Invitrogen
Manufacturer Code:MA181733
Catalog # PIMA181733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | H202-106 |
Immunogen | MTE1/MTE2 stromal cell lines. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: BV11 antigen; CD321; CD321 JAM JAM1 JAMA JCAM KAT PAM-1 F11R; F11 receptor/F11R; JAM-1; JAM-A; junction cell adhesion molecule A; junction cell adhesion molecule1; Junctional adhesion molecule 1; Junctional adhesion molecule A
Gene Aliases: 9130004G24; AA638916; ESTM33; F11r; JAM; JAM-1; JAM-A; Jam1; Jcam; Jcam1; Ly106
UniProt ID: (Mouse) O88792
Entrez Gene ID: (Mouse) 16456
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.