Abnova
Manufacturer Code:H00003718M04A
Catalog # 89-104-795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D5 |
Immunogen | JAK3 (AAH280681 a.a. &bsim 619 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant JAK3.
For Research Use Only
Protein Aliases: JAK-3; JAK3_HUMAN; JAKL; Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); L-JAK; Leukocyte janus kinase; LJAK; Tyrosine-protein kinase JAK3
Gene Aliases: JAK-3; JAK3; JAK3_HUMAN; JAKL; L-JAK; LJAK
UniProt ID: (Human) P52333
Entrez Gene ID: (Human) 3718
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.