Invitrogen
Manufacturer Code:PA5106248
Catalog # PIPA5106248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human JAK2/3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Breast-JAK3 fae Fd17 Jak2 JAK-2 Jak3 JAK-3 JAK3_HUMAN JAK3B JAKL Janus kinase 2 Janus kinase 2 (a protein tyrosine kinase) Janus kinase 3 Janus kinase 3 (a protein tyrosine kinase leukocyte) Janus kinase 3 protein-tyrosine kinase Janus kinase 3 protein-tyrosine kinase JTK10 Leukocyte janus kinase LJAK L-JAK RATJAK3 THCYT3 Tyrosine-protein kinase JAK2 tyrosine-protein kinase JAK3 wil; JAK-2; JAK-3; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); Janus kinase 3 protein-tyrosine kinase; Janus kinase 3, protein-tyrosine kinase; L-JAK; Leukocyte janus kinase; Tyrosine-protein kinase JAK2; Tyrosine-protein kinase JAK3
Gene Aliases: fae; Fd17; JAK-3; JAK2; JAK3; JAK3_HUMAN; JAKL; JTK10; L-JAK; LJAK; RATJAK3; THCYT3; wil
UniProt ID: (Human) O60674, (Human) P52333, (Rat) Q62689, (Mouse) Q62120, (Mouse) Q62137, (Rat) Q63272
Entrez Gene ID: (Human) 3717, (Human) 3718, (Rat) 24514, (Mouse) 16452, (Mouse) 16453, (Rat) 25326
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.