Invitrogen
Manufacturer Code:PA5109871
Catalog # PIPA5109871
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 120-380 of human JAK1 (NP_002218.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA960307 BAP004 C130039L05Rik EC 2.7.10.2 JAK1 JAK-1 JAK1A JAK1B Janus kinase 1 JTK3 kinase Jak1 Tyrosine-protein kinase JAK1; JAK-1; Janus kinase 1; Tyrosine-protein kinase JAK1
Gene Aliases: AA960307; BAP004; C130039L05Rik; JAK1; JAK1A; JAK1B; JTK3
UniProt ID: (Human) P23458
Entrez Gene ID: (Human) 3716, (Mouse) 16451
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.