Invitrogen
Manufacturer Code:PA572883
Catalog # PIPA572883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | JAGN1 antibody for a 19 amino acid peptide near the carboxy terminus of human JAGN1. The immunogen is within the last 50 amino acids of JAGN1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein jagunal homolog 1
Gene Aliases: 5830427H10Rik; GL009; JAGN1; SCN6
UniProt ID: (Human) Q8N5M9, (Mouse) Q5XKN4, (Rat) Q4KM64
Entrez Gene ID: (Human) 84522, (Mouse) 67767, (Rat) 502872
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.