Invitrogen
Manufacturer Code:MA123248
Catalog # PIMA123248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8H8.5 |
Immunogen | The full-length mouse JAB-1 gene expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 38 kDa Mov34 homolog; COP9 complex S5; COP9 constitutive photomorphogenic homolog subunit 5; COP9 signalosome complex subunit 5; CSN5 JAB1 MGC3149 MOV-34 SGN5; Jun activation domain-binding protein 1; Jun coactivator; Kip1 C-terminus-interacting protein 2; SGN5; signalosome subunit 5; testis secretory sperm-binding protein Li 231m
Gene Aliases: AI303502; COPS5; CSN5; JAB1; Kic2; MOV-34; Mov34; SGN5
UniProt ID: (Human) Q92905, (Mouse) O35864
Entrez Gene ID: (Human) 10987, (Mouse) 26754
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.