Novus Biologicals
Manufacturer Code:NBP184370
Catalog # NBP184370
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ANNMQEAQSIDEIYKYDKKQQQEILAAKPWTKDHHYFKYCKISALALLKMVMHARSGGNLEVMGLMLGKVDGETMIIMDSFALPVEGTETRVNAQAAAYEYMAAYIENAKQVGRLENAIGWYHSHPGYGCWLS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 38 kDa Mov34 homolog; COP9 constitutive photomorphogenic homolog subunit 5; COP9 constitutive photomorphogenic homolog subunit 5 (Arabidopsis) COP9 signalosome complex subunit 5 CSN5COP9 (constitutive photomorphogenic Arabidopsis homolog) subunit 5 EC 3.4 JAB138 kDa Mov34 homolog Jun activation domain-binding protein 1 MOV-34 SGN5MGC3149 Signalosome subunit 5; COP9 signalosome complex subunit 5; Jun activation domain-binding protein 1; SGN5; signalosome subunit 5; testis secretory sperm-binding protein Li 231m
Gene Aliases: COPS5; CSN5; JAB1; MOV-34; SGN5
UniProt ID: (Human) Q92905
Entrez Gene ID: (Human) 10987
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.