Proteintech
Manufacturer Code:275111AP150UL
Catalog # 50-173-5689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | JAB1 Fusion Protein Ag25950 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 38 kDa Mov34 homolog; COP9 constitutive photomorphogenic homolog subunit 5; COP9 signalosome complex subunit 5; COPS5 CSN5 JAB1 MOV 34 SGN5 Signalosome subunit 5; Jun activation domain-binding protein 1; SGN5; signalosome subunit 5; testis secretory sperm-binding protein Li 231m
Gene Aliases: COPS5; CSN5; JAB1; MOV-34; SGN5
UniProt ID: (Human) Q92905
Entrez Gene ID: (Human) 10987
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.