Abnova
Manufacturer Code:MAB5519
Catalog # 89-112-274
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | F11 |
Immunogen | Native purified Itgb3 from newborn rat bone cell suspension. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store in the dark at 4°C. Do not freeze.<br />Avoid prolonged exposure to light.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against native Itgb3.
For Research Use Only
Protein Aliases: GPIIIa; integrin beta 3 (Cd61); Integrin beta-3; Platelet membrane glycoprotein IIIa
Gene Aliases: Itgb3
UniProt ID: (Rat) Q8R2H2
Entrez Gene ID: (Rat) 29302
Molecular Function:
cell adhesion molecule
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.