Abnova
Manufacturer Code:MAB10946
Catalog # 89-109-159
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RMV-7 |
Immunogen | Mouse Itgav. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rat monoclonal antibody raised against Itgav.
For Research Use Only
Protein Aliases: 1110004F14Rik; 2610028E01Rik; alpha v integrin; CD51; D430040G12Rik; Integrin alpha-V; Integrin alpha-V heavy chain; Integrin alpha-V light chain; vitronectin receptor alpha polypeptide (VNRA); Vitronectin receptor subunit alpha
Gene Aliases: 1110004F14Rik; 2610028E01Rik; CD51; D430040G12Rik; Itgav
UniProt ID: (Mouse) P43406
Entrez Gene ID: (Mouse) 16410
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.