Abnova
Manufacturer Code:MAB5780
Catalog # 89-112-451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | DX5 |
Immunogen | RIL-2-propagated NK1.1+ cells from C57BL/6 mice. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store in the dark at 4°C. Do not freeze.<br />Avoid prolonged exposure to light.<br />Aliquot to avoid repeated freezing and thawing. |
Rat monoclonal antibody raised against Itga2.
For Research Use Only
Protein Aliases: CD49 antigen-like family member B; CD49B; Collagen receptor; DX5; GPIa; Integrin alpha-2; Platelet membrane glycoprotein Ia; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha
Gene Aliases: CD49B; DX5; Itga2
UniProt ID: (Mouse) Q62469
Entrez Gene ID: (Mouse) 16398
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.