Novus Biologicals
Manufacturer Code:NBP270988AF350
Catalog # NP270988A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2H9 |
Immunogen | Full-length protein expressed in 293T cell transfected with human Isocitrate Dehydrogenase 1/IDH1 expression vector. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Cytosolic NADP-isocitrate dehydrogenase; Cytosolic NADP-isocitrate dehydrogenase EC 1.1.1.42 IDCD IDH IDP IDPC isocitrate dehydrogenase [NADP] cytoplasmic isocitrate dehydrogenase 1 (NADP&plus) soluble NADP(&plus)-specific ICDH NADP-dependent isocitrate dehydrogenase cytosolic NADP-dependent isocitrate dehydrogenase peroxisomal Oxalosuccinate decarboxylase PICD; epididymis luminal protein 216; epididymis secretory protein Li 26; IDH; IDP; isocitrate dehydrogenase 1 (NADP+); isocitrate dehydrogenase 1 (NADP+), soluble; Isocitrate dehydrogenase [NADP] cytoplasmic; NADP(+)-specific ICDH; NADP-dependent isocitrate dehydrogenase, cytosolic; NADP-dependent isocitrate dehydrogenase, peroxisomal; Oxalosuccinate decarboxylase
Gene Aliases: HEL-216; HEL-S-26; IDCD; IDH; IDH1; IDP; IDPC; PICD
UniProt ID: (Human) O75874
Entrez Gene ID: (Human) 3417
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.